Drukuj
Nauszniki, Peltor™ X Series
Dostawca: 3M
X5A X5P3E X1P3E X2A X2P3E X3A X3P3E X4A X4P3E X1A
111-1139EA 372 PLN
111-1139 111-1141 111-1132 111-1133 111-1134 111-1135 111-1136 111-1137 111-1138 111-1131
Nauszniki, Peltor™ X Series
Nauszniki Nałuszniki tłumiące hałas
These earmuffs combine excellent protection with ultimate comfort. They are available as headband and helmet-mounted versions, offering a wide range of attenuation levels that meet the needs of the majority of industrial applications.
  • Soft, wide cushions reduce pressure around the ears and improve comfort and wearability
  • Slimline, dual-moulded cups
  • Large space inside cups reduces moisture and heat build-up
  • Electrically-insulated wire headband
  • Colour coded cups: Easy to identify level of attenuation
X1: Ultra-slim earmuffs for low noise environments.
X2: Lightweight earmuffs for low to moderate noise environments.
X3: High protection earmuffs for moderate to high noise environments.
X4: High level of protection without all the bulk, the innovative foam earcup inserts and spacers help boost attenuation while adding little bulk or weight.
X5: For very high noise situations, high attenuation, twin headband design helps reduce heat build-up with good fit and balance.
Suitable applications:
X1A / X1P3: Lawn mowing, power drilling
X2A / X2P3: Construction, manufacturing industry
X3A / X3P3: Forestry, industry, airports, engineering
X4A / X4P3: Industry
X5A / X5P3: Mining, quarrying, paper manufacturing, gas turbines.
Dodatkowe informacje: Hygiene kits contain replacement sealing rings and muffler pads which can be replaced in a few easy steps. It extends the life of earmuffs and ensures good function and hygiene.