Drukuj
Kolumienki SPE, CHROMABOND® HR-X
Dostawca: MACHEREY-NAGEL
730938 730931MPS 730931 730939 730934 730936 730935 730937 730940 730941 738530.050M 738530.025M 738530.100M 730931.250 730931P45 730934P45 730935MPS 730935P45 730936P45 730938.250 730939.250 730939MPS 732132
705-0771EA 735 PLN
705-0771 MANA730931MPS 705-0769 705-0772 705-0766 705-0768 705-0767 705-0770 705-0773 705-0774 MANA738530.050M MANA738530.025M MANA738530.100M MANA730931.250 MANA730931P45 MANA730934P45 MANA730935MPS MANA730935P45 MANA730936P45 MANA730938.250 MANA730939.250 MANA730939MPS MANA732132
Kolumienki SPE, CHROMABOND® HR-X
Kolumny chromatograficzne
CHROMABOND® HR-X is a spherical, hydrophobic polystyrene-divinylbenzene resin for SPE.
  • Applicable for a wide range of analytes
  • Excellent recovery rates due to the spherical structure of the particles
  • High-purity material with high reproducibility
Applications: Active ingredients from tablets, creams and water; drugs and pharmaceuticals from urine, blood serum and plasma; trace analysis of pesticides, phenols, PAHs and PCBs from water etc.