Drukuj
Wagi przemysłowe, kompaktowe wagi stołowe Ranger® 3000
Dostawca: OHAUS
30060902 30031703 30031706 30031704 30031705 30060904 30060905 30060906 30031702 30060903
611-2596EA 1955 PLN
611-2596 611-2582 611-2585 611-2583 611-2584 611-2598 611-2599 611-2611 611-2581 611-2597
Wagi przemysłowe, kompaktowe wagi stołowe Ranger® 3000
Wagi
Simple to operate, multipurpose, compact scale for producing precise results rapidly. Application modes include weighing, percent weighing, parts counting, accumulation and dynamic weighing.
  • Large backlit LCD
  • Portable with ABS lightweight plastic housing (net weight 4,2 kg) and rechargeable battery
  • Simple keyboard, five-key operation
  • Stabilisation within 1 second
  • Weigh below capability
Informacje do zamówienia: More connectivity options available, including optional Ethernet, 2nd RS232 or USB and optional 2nd platform. Approved versions are also available. Please see the website or contact your local Avantor sales office for more details.