Drukuj
Wagi przemysłowe, kompaktowe wagi stołowe Valor™ 7000
Dostawca: OHAUS
30060916 30060912 30031823 30031824 30031826 30060915 30031825 30060913 30031822 30060914
611-2621EA 2275 PLN
611-2621 611-2617 611-2592 611-2593 611-2595 611-2620 611-2594 611-2618 611-2591 611-2619
Wagi przemysłowe, kompaktowe wagi stołowe Valor™ 7000
Wagi
A food scale designed to increase productivity of food weighing and proportioning tasks, that incorporates touchless sensors to operate certain functions without the touch of a button, thereby reducing scale contamination. Valor 7000 has Food Safety Certification (NSF) and supports a HACCP compliant quality system.
  • Front and rear touchless sensors
  • Dual backlit displays, front and rear for quick weighing by two operators
  • One second (1 s) stabilisation time for increased throughput
  • Mains supply with internal rechargeable battery for portable use
  • IP 43 (splash) protection
Informacje do zamówienia: More connectivity options available, including optional Ethernet, second RS232 or USB. Approved versions are also available. Please see the website or enquire for more details.