Drukuj
Wkłady SPE, (Na2SO4) CHROMAFIX® Dry
Dostawca: MACHEREY-NAGEL
731852 731853 731854
MANA731852EA 615 PLN
MANA731852 MANA731853 MANA731854
Wkłady SPE, (Na2SO4) CHROMAFIX® Dry
Kolumny chromatograficzne
CHROMAFIX® Dry is a special phase for drying organic samples.
  • Polypropylene cartridges in three different sizes with different adsorbent weights (S: 0,4 ml, M: 0,8 ml, L: 1,8 ml)
  • Anhydrous high-purity sodium sulfate which forms Glauber’s salt with traces of water
Application: Connected with a syringe via Luer fittings traces of water from organic solutions will be removed. For removal of larger quantities of water several cartridges can be combined in series.