Drukuj
Kolumienki SPE, CHROMABOND® HR-P
Dostawca: MACHEREY-NAGEL
730108 730108G 730111 730111G 730117 730118 730119 730280 731839 731840 731841 732108 738111.100M 730108.250
554-6594EA 552 PLN
554-6594 MANA730108G MANA730111 MANA730111G MANA730117 MANA730118 MANA730119 MANA730280 MANA731839 MANA731840 MANA731841 MANA732108 MANA738111.100M MANA730108.250
Kolumienki SPE, CHROMABOND® HR-P
Kolumny chromatograficzne
CHROMABOND® HR-P is a polystyrene-divinylbenzene adsorbent resin for SPE.
  • Highly porous copolymer with a very large surface
  • Very high binding capacity compared with silica adsorbents
Recommended for aromatic compounds, like phenols, nitroaromatics, and pesticides from water or PAHs from oil.