Drukuj
Kolumienki SPE, CHROMABOND® Florisil®
Dostawca: MACHEREY-NAGEL
730081 730082 730082G 730238 730457 731848 730239 730082.250 730238G 730239G
MANA730081EA 749 PLN
MANA730081 MANA730082 MANA730082G MANA730238 MANA730457 MANA731848 MANA730239 MANA730082.250 MANA730238G MANA730239G
Kolumienki SPE, CHROMABOND® Florisil®
Kolumny chromatograficzne
CHROMABOND® Florisil® is a magnesium silicate for SPE.
  • High purity magnesium silicate (MgO – SiOH 15:85)
  • Polar phase
  • Alternative selectivity to SiOH
Recommended for polar and aromatic substances like organic tin compounds, aliphatic carboxylic acids, PCBs, and PAHs.