Drukuj
Probówki pediatryczne +E17959:E18073 z zakrętką, Vacuette®
Dostawca: Greiner Bio-One
459036_100 459075_100 459084_100 459085_100
GREI459036_100EA 219 PLN
GREI459036_100 GREI459075_100 GREI459084_100 GREI459085_100
Probówki pediatryczne +E17959:E18073 z zakrętką, Vacuette®
Systemy do diagnostyki klinicznej Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clar.
The innovative VACUETTE® blood collection tubes made from practically unbreakable plastic have become well established on the market. They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.