Hand-Operated Piston Dispensing Drum Pump, PVC, Spout Outlet, 16 Strokes/gallon
Dostawca: Avantor Fluid Handling
Economical piston pumps are ideal for a variety of fluids.
- Adjustable to most pails and drums
Models 07079-50 and -55 feature an angled spout to help prevent dripping. Models 07079-60 and -65 are made entirely of PVC, including the piston and seal, making them an ideal option for sanitary applications.