Drukuj
Masterflex® Transfer Tubing, Metric PTFE, 7.98 mm ID x 10.0 mm OD; 25 ft
Dostawca: Avantor Fluid Handling
VWR Item#MFLX06605-56
Your Organization's Price:
Withstands extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 7.98 mm ID x 10.0 mm OD
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. <br />Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. <br />Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. <br />Temperature range: -400 to 500 °F (-240 to 260 °C) <br />Sterilize: With autoclave, ethylene oxide, or use dry heat.