Masterflex® L/S® Bulk-Packed High-Performance Precision Tubing, C-Flex®, L/S 24; 20 Bags of 25 ft/Bg
Dostawca: Avantor Fluid Handling
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Very low protein binding
- Heat sealable, weldable
- Economical tubing
- Tubing Size: L/S 24
<B>Applications</B>: General-purpose lab uses requiring high performance and long life<br /><B>Characteristics</B>: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white<br />Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.