Masterflex® Transfer Tubing, Metric PTFE, 10.0 mm ID x 12.0 mm OD; 25 ft
Dostawca: Avantor Fluid Handling
Withstands extreme temperatures.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Tubing size: 10.0 mm ID x 12.0 mm OD
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. <br />Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. <br />Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. <br />Temperature range: -400 to 500 °F (-240 to 260 °C) <br />Sterilize: With autoclave, ethylene oxide, or use dry heat.