Drukuj
Skrobaki do komórek
Dostawca: witeg Labortechnik
8.486.200
WITG8.486.200EA 477 PLN
WITG8.486.200
Skrobaki do komórek
Skrobaki do komórek
Bent, plastic coated glass handle, mobile PTFE blade. Scraper for removal of cells from flasks and plates. Length 50 mm.