Drukuj
Probówki do NMR, cienkościenne, PRECISION, 3 mm Ø, SP Wilmad-LabGlass
Dostawca: SP Industries
335-PP-9-5 327-PP-7-5 328-PP-8-5 335-PP-8-5 328-PP-7-5 341-PP-7-5 307-PP-7-5
WILM335-PP-9-5EA 1130 PLN
WILM335-PP-9-5 WILM327-PP-7-5 WILM328-PP-8-5 WILM335-PP-8-5 WILM328-PP-7-5 WILM341-PP-7-5 WILM307-PP-7-5
Probówki do NMR, cienkościenne, PRECISION, 3 mm Ø, SP Wilmad-LabGlass
Probówki Probówki do NMR
Borosilicate glass, Class A, ASTM Type 1.
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
  • To maximise SNR, Precision NMR tubes have minimal paramagnetic impurities that would impact shimming
  • Tight int.Ø and Ø tolerance as small as 0,0065 mm accommodates inserts
  • Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6
NMR PRECISION tubes are manufactured from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438. PRECISION tube has 3 fold less paramagneticWięcej informacji o produkcie