Drukuj
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
Dostawca: DWK Life Sciences
W988003-S W988004-S W988005-S W988006-S W988007-S W988008-S
WHEAW988003-SEA 6000 PLN
WHEAW988003-S WHEAW988004-S WHEAW988005-S WHEAW988006-S WHEAW988007-S WHEAW988008-S
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
Korki Korki gumowe
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-use, Just-in-time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
Certyfikaty: USP 788 / EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP 85/ EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP 660 / EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381 / EP 3.2.9Więcej informacji o produkcie