Drukuj
Filtry Whatman™ Polydisc™ AS In-Line, produkty Whatman (Cytiva)
Dostawca: Whatman products (Cytiva)
6724-5002 6724-5145
516-1108EA 1140 PLN
516-1108 WHAT6724-5145
Filtry Whatman™ Polydisc™ AS In-Line, produkty Whatman (Cytiva)
Filtry Filtry przepływowe (in-line)
Whatman Polydisc AS (Aqueous Solution) in-line filters from Cytiva feature high-throughput PES membrane with a glass microfiber prefilter. Polydisc comes presterilized in medical-grade blister packs
  • Whatman filters with polyethersulfone membranes have low protein binding and are surfactant-free
  • GMF prefilter removes heavy particulates, increasing membrane longevity
  • Radiation sterilized, so there are no ethylene oxide (EtO) residuals
  • Barbed hose connections fit multiple tubing sizes
  • Integrity-testable with the bubble point method
  • Lightweight design prevents
Więcej informacji o produkcie