Drukuj
Schodki bezpieczne
Dostawca: WAKU GERATE
210/C 310/C 510/B 410B 310B 220C
139-9935EA 2350 PLN
139-9935 139-9936 139-9938 WAKU410B WAKU310B WAKU220C
Schodki bezpieczne
Drabiny i schodki
Light, aluminium safety steps. Strong double rails with robust, tension and pressure resistant bracings for increased stability. Platform height corresponds to height of top step.
  • Anti-slip plastic feet
  • The large treads (36×23 cm) are covered with a non-slip rubber lining.
TÜV tested (DIN 4566 and 4567).
Certyfikaty: TÜV tested (DIN 4566 and 4567).