Drukuj
PYROSTAR™ Neo+ Endotoxin Detection Reagent
Dostawca: FUJIFILM WAKO CHEMICALS
293-36941
WAKF293-36941EA 2040 PLN
WAKF293-36941
PYROSTAR™ Neo+ Endotoxin Detection Reagent
Systemy do testów mikrobiologicznych Testy endotoksyn
PYROSTAR Neo+ is a new endotoxin detection reagent developed by recombinant technology.
  • Colorimetric method, can be used with absorption plate reader
  • Same cascade reaction as Lysate reagent, conforms to traditional detection
  • Endotoxin specific reagent, no false positive risk of (1→3)-β-D-glucan
  • Determination range from 0,001 to 50 EU/ml
  • High sensitivity, reagent reacts with endotoxin over a wide range
Combine with traditional lysate reagent and recombinant 3 factors (factor C, factor B, Proclotting Enzyme) from Limilus polyphemus. It is a high sensitive colorimetric assay specific for endotoxin.