Drukuj
Limulus Test Tube
Dostawca: FUJIFILM WAKO CHEMICALS
292-32751 293-26551
WAKF292-32751EA 1400 PLN
WAKF292-32751 WAKF293-26551
Limulus Test Tube
Probówki Probówki
Our specialised Limulus Test Tube-S are made of high-quality borosilicate glass depyrogenated at 250 °C.
  • Endotoxin-free test tubes (<0,001 EU/tube)
  • Designed for gel clot, KTA and KCA testing on the Toxinometer®
  • Made with borosilicate glass
These tubes have a special curvature that allows the appropriate amount of light to transmit through the sample to measure the endotoxin concentration based on the method used. This glass tube is designed for the Toxinometer® Measurement System.
The Depyrogenated Aluminum Caps assists in reducing the chance ofWięcej informacji o produkcie