Drukuj
Komfortowy zestaw kartridżowy, arium®
Dostawca: Sartorius
H2O-C-PACK
VIVAH2O-C-PACKEA 2520 PLN
VIVAH2O-C-PACK
Komfortowy zestaw kartridżowy, arium®
Filtry Wkłady filtracyjne
This is the deionization cartridge set with top-down technology which is optimized for the removal of both organic and inorganic constituents. It is specifically designed to match the unit and delivers ultrapure water which exceeds the ASTM Type 1 quality standard.
  • Long lasting high performance and low maintenance operation
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • Optimised crossflow behavior, prevents separation of the resin mixed-bed
  • Patented connection process simplifies the replacement of consumables