Drukuj
Rurki, Tygon® 3370 I.B.
Dostawca: Saint Gobain Life Sciences
T1316-63 T1312-43 T1308-43 T1306-43 T1320-63 T1324-63 T1332-63 T1340-66
VERNT1316-63EA 11720 PLN
VERNT1316-63 VERNT1312-43 VERNT1308-43 VERNT1306-43 VERNT1320-63 VERNT1324-63 VERNT1332-63 VERNT1340-66
Rurki, Tygon® 3370 I.B.
Przewody, kapilary i rury Przewody rurowe
Braid reinforced for increased pressure resistance, TYGON® 3370 I.B. silicone hose is frequently specified in the most demanding applications requiring sanitary transfer of fluids.
  • Consistently smooth inner surface limits particle entrapment
  • Platinum cured to minimize extractables
  • Tough braid reinforcement permits use under elevated working pressures
  • Withstands repeated CIP and SIP cleaning and sterilization
Its smooth inner surface reduces the risk of particle entrapment and inhibits excessive residue and microscopic bacterial build-up; cleaning andWięcej informacji o produkcie