Drukuj
Węże Versilon™ C-210-A
Dostawca: Saint Gobain Life Sciences
AEM02044 AEM02046 AEM02053 AEM02062 AEM02002 AEM02006 AEM02007 AEM02011 AEM02012 AEM02013 AEM02016 AEM02017 AEM02018 AEM02019 AEM02022 AEM02027 AEM02028 AEM02029 AEM02034 AEM02037 AEM02038
VERNAEM02044EA 3900 PLN
VERNAEM02044 VERNAEM02046 VERNAEM02053 VERNAEM02062 VERNAEM02002 VERNAEM02006 VERNAEM02007 VERNAEM02011 VERNAEM02012 VERNAEM02013 VERNAEM02016 VERNAEM02017 VERNAEM02018 VERNAEM02019 VERNAEM02022 VERNAEM02027 VERNAEM02028 VERNAEM02029 VERNAEM02034 VERNAEM02037 VERNAEM02038
Węże Versilon™ C-210-A
Przewody, kapilary i rury Przewody rurowe
Polyurethane, formulation C-210-A, translucent.
  • Consistently tight dimensional tolerance
  • Excellent abrasion and tear resistance
  • Excellent resistance to oils, greases and fuels
  • Retains flexibility in sub-zero environments
  • High tear resistance
A strictly controlled manufacturing process ensures this hose has a consistently low dimensional tolerance. This combined with high elasticity means the tubing can be connected to fittings easily and without any problems.
Temperature resistance: −73 to +93 °C
Hardness (Shore durometer A): 82