Drukuj
Cleandox® Cleanroom Papers
Dostawca: VELTEK ASSOCIATES
C2WR-CDA4-01 C2WRCD8.5X1101 C2WRCD85X11BL0 C2WRCD85X11CR0 C2WRCD85X11GR0 C2WRCD85X11PK0 C2WRCDA4BLU01 C2WRCDA4CRM01 C2WRCDA4GRE01 C2WRCDA4PNK01
VELTC2WR-CDA4-01EA 1500 PLN
VELTC2WR-CDA4-01 VELTC2WRCD8.5X1101 VELTC2WRCD85X11BL0 VELTC2WRCD85X11CR0 VELTC2WRCD85X11GR0 VELTC2WRCD85X11PK0 VELTC2WRCDA4BLU01 VELTC2WRCDA4CRM01 VELTC2WRCDA4GRE01 VELTC2WRCDA4PNK01
Cleandox® Cleanroom Papers
Artykuły papiernicze
CleanDox® is a poly-infused bond paper that can be used throughout your facility for note taking, printing batch records, forms and logbooks.
  • Non-sterile and ISO 5 Cleanroom processed
  • Vacuum packed with double layers cleanroom bag, packed in controlled ISO 5 cleanroom environment
  • Low in chloride and other ionic contamination
  • Formulated without magnesium silicate
  • Excellent IPA resistant material
  • Compatible with most inks used for writing without smearing
  • Ultra low ionic contamination, ultra low particle and fiber generation
  • Excellent heat resistant material
Więcej informacji o produkcie