Drukuj

Anti-TP53 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: DO-1]

Dostawca: US Biological
P1001-26U1
USBIP1001-26U1EA 2620 PLN
USBIP1001-26U1
Anti-TP53 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: DO-1]
Przeciwciała
Recommended dilution: Flow Cytometry: 1:2-1:10;

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: DO-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR