Drukuj
Zestawy 1-etapowe ExoProStar™ illustra™
Dostawca: Cytiva
US77702 US77705 US77720 US77750 US77701
US77702EA 619 PLN
US77702 US77705 US77720 US77750 US77701
Zestawy 1-etapowe ExoProStar™ illustra™
Odczynniki do kwasów nukleinowych Odczynniki i zestawy do oczyszczania kwasów nukleinowych
illustra™ ExoProStar™ 1-STEP Kit uses illustra™ Exonuclease I and Alkaline Phosphatase for optimal removal of unincorporated primers and nucleotides. illustra™ ExoProStar™ improves digestion efficiency with no degradation of the target PCR product. The sample remains completely intact and ready for immediate use in manual or automated processes.
  • Both enzymes provided in one tube with just one simple pipetting step needed to prepare the reaction
  • Fast 30 minute protocol - complete heat inactivation of the enzymes within 15 minutes
  • Adaptable to different sizes
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate