Drukuj
Żywice do chromatografii oddziaływań hydrofobowych, Toyopearl®, TSK-GEL®
Dostawca: TOSOH Bioscience
0007476 0007477 0007478 0014477 0014478 0014479 0014701 0014718 0014720 0016050 0016052 0016172 0016173 0019026 0043151 0043152 0043153 0043176 0043177 0043276 0043277 0019818 0044465 0014703 0019956 0021448 0021888 0043126 0019027 0019955 0019957 0016174 0014783 0014784 0014785 0016175 0043127 0021449 0021450 0021889 0021887 0019802 0021301
TOYO0007476EA 3550 PLN
TOYO0007476 TOYO0007477 TOYO0007478 TOYO0014477 TOYO0014478 TOYO0014479 TOYO0014701 TOYO0014718 TOYO0014720 TOYO0016050 TOYO0016052 TOYO0016172 TOYO0016173 TOYO0019026 TOYO0043151 TOYO0043152 TOYO0043153 TOYO0043176 TOYO0043177 TOYO0043276 TOYO0043277 TOYO0019818 TOYO0044465 TOYO0014703 TOYO0019956 TOYO0021448 TOYO0021888 TOYO0043126 TOYO0019027 TOYO0019955 TOYO0019957 TOYO0016174 TOYO0014783 TOYO0014784 TOYO0014785 TOYO0016175 TOYO0043127 TOYO0021449 TOYO0021450 TOYO0021889 TOYO0021887 TOYO0019802 TOYO0021301
Żywice do chromatografii oddziaływań hydrofobowych, Toyopearl®, TSK-GEL®
Sorbenty chromatograficzne Sorbenty do HPLC
Hydrophobic interaction chromatography (HIC) is a powerful tool for the process purification of biomolecules. Five HIC ligands featuring different degrees of hydrophobicity and selectivity. The hydrophobicity of HIC resins increases through the ligand series.
  • Typically used in intermediate purification and polishing steps
Ether HIC resins: Ether hydrophobic interaction chromatography (HIC) resins have the highest hydrophilicity (lowest hydrophobicity) of the HIC ligands. The ether ligand is useful for very hydrophobic molecules such as membrane proteins. It is typically used in intermediate purification and polishing steps.
Phenyl HIC resins: Phenyl hydrophobic interaction chromatography resins are in the middle of the hydrophobicity range. Primary applications are for the separation of proteins, their isoforms, and aggregate removal.
Butyl HIC resins: Butyl hydrophobic interaction chromatography (HIC) resins have the second highest hydrophobicity of the HIC ligands. There are three pore sizes available for this ligand and each has been optimised for a specific molecular size profile. The primary applications are the separation of proteins, their isoforms, and aggregate removal.
Hexyl HIC resins: Hexyl hydrophobic interaction chromatography (HIC) resin has the highest hydrophobicity of the HIC ligands. It is used for very hydrophilic proteins or where a low salt elution environment is required.