Drukuj
Hygienox Electropolished Seating Benches
Dostawca: TEKNOMEK
CRSEHYEPSB1035 CRSEHYEPSB1235 CRSEHYEPSB1535 CRSEHYEPSB1835 CRSEHYEPSB2035 CRSEHYEPSB2535
TEKMCRSEHYEPSB1035EA 0 PLN
TEKMCRSEHYEPSB1035 TEKMCRSEHYEPSB1235 TEKMCRSEHYEPSB1535 TEKMCRSEHYEPSB1835 TEKMCRSEHYEPSB2035 TEKMCRSEHYEPSB2535
Hygienox Electropolished Seating Benches
Meble Stoły
Hygienox electropolished 304L-grade stainless steel seating benches are made specifically for cleanroom environments, giving extreme protection against pitting and crevice corrosion caused by harsh clean down routines.
  • Optimum sitting height
  • Easy to swivel over
  • Four stainless steel adjustable feet with non-marking rubber base
  • Use a joining bracket to create a larger bench
Providing protection from corrosive chemicals, such as chlorine and hydrogen peroxide, the hygienox seating benches are also easier to clean due to the smoother surface of theWięcej informacji o produkcie