Drukuj
Alkometr, rekalibracja
Dostawca: SURESCREEN
AMCAL
SUREAMCALEA 652 PLN
SUREAMCAL
Alkometr, rekalibracja
Testy narkotykowe
After 500 uses the alcometer requires recalibration to ensure that the instrument is reading correctly. It is returned to manufacturer for this service, and turn around is five working days from receipt.