Drukuj
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Dostawca: Merck
39361-250ML
SUPL39361-250MLEA 267 PLN
SUPL39361-250ML
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Standardy Wzorce kationowe Silver Standards
This silver standard solution prepared with high quality silver metal in 2% HNO3.
Certyfikaty: ISO/IEC 17025 and ISO Guide 34. Traceable to NIST, SRM.