Drukuj
Spektrofotometry UV, seria evolution
Dostawca: Thermo Fisher Scientific
840-370100 840-372600 840-372700 912A1111 840-340200 840-370200 840-370400 840-370300 840-341500 840-340100 912A1110 840-370500 912A1112 840-370700 840-370600 840-370800
SPET840-370100EA 94820 PLN
SPET840-370100 SPET840-372600 SPET840-372700 SPET912A1111 SPET840-340200 SPET840-370200 SPET840-370400 SPET840-370300 SPET840-341500 SPET840-340100 SPET912A1110 SPET840-370500 SPET912A1112 SPET840-370700 SPET840-370600 SPET840-370800
Spektrofotometry UV, seria evolution
Spektrofotometry i fotometry Spektrofotometry UV/Vis
Empower the analysis and bring versatility and productivity to your lab with the evolution uv-vis spectrophotometers. With next-generation INSIGHT™ PRO software, the instrument is always up to date and ready for the next sample challenge.
  • Double-beam geometry
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices to complete the analysis
For routine to research analysis, this instrument series offers real innovationsWięcej informacji o produkcie