Drukuj
Tygle filtracyjne, DURAN®
Dostawca: DWK Life Sciences
258510203 258511105 258511208 258511302 258510306 258512101 258513106 258512504 258510409 258511405 258512204 258512401 258513209 258513303 258513406 258513509 258512307
SCOT258510203EA 911 PLN
SCOT258510203 SCOT258511105 SCOT258511208 SCOT258511302 SCOT258510306 SCOT258512101 SCOT258513106 SCOT258512504 SCOT258510409 SCOT258511405 SCOT258512204 SCOT258512401 SCOT258513209 SCOT258513303 SCOT258513406 SCOT258513509 SCOT258512307
Tygle filtracyjne, DURAN®
Tygle
These filtration crucibles are made from clear DURAN® borosilicate glass 3.3.
  • Good thermal shock and chemical resistance
Typical applications include qualitative inorganic analysis and preparative chemistry.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable