Drukuj
Łaźnie termostatyczne do wiskozymetrów, CT 72
Dostawca: SI Analytics
285418526 285418547 285418568 285418501 285418615
SCOI285418526EA 32920 PLN
SCOI285418526 SCOI285418547 645-0081 645-0080 SCOI285418615
Łaźnie termostatyczne do wiskozymetrów, CT 72
Łaźnie Łaźnie wiskozymetryczne
The CT 72 thermostatic baths are designed for determining viscosity of newtonian liquids in glass capillary viscometers with the display of the momentary and the set temperature through VF display.
  • User friendly through automatic fuses on the back panel instead of micro fuses
  • Programmable set temperatures through integrated clock with program controller
  • Easy to reach overtemperature safety system on front panel