Drukuj
Piec Headspace i podajnik próbek do kulometrycznego miareczkowania KF
Dostawca: SI Analytics
285227850 285227820 285227840
SCOI285227850EA 122610 PLN
SCOI285227850 SCOI285227820 SCOI285227840
Piec Headspace i podajnik próbek do kulometrycznego miareczkowania KF
Titratory
The headspace oven technique of the TO 7280 allows the separation of the water to be titrated from liquid, solid and semi-solid (paste-like) samples. In combination with the TW 7650 sample changer, up to 49 samples can be analysed.
  • Semi-automatic or fully automatic water determination
  • No contamination of the anolyte, generator and indicator electrodes
  • Application range from 35 to 280 °C
  • Reproducibility: ±3 μg bei 10 to 1000 μg, 0,3% bei >1 mg
Requires TitriSoft version 3.50 or higher to operate. One position is reserved for the zero-vial toWięcej informacji o produkcie