Drukuj
ThermoSafe® ChillTech® Pre-Qualified 2 to 8 °C PCM Shippers
Dostawca: Sonoco Thermosafe
CHR20U CHR5U
SCATCHR20UEA 2150 PLN
SCATCHR20U SCATCHR5U
ThermoSafe® ChillTech® Pre-Qualified 2 to 8 °C PCM Shippers
Koperty i pojemniki wysyłkowe
ChillTech® is a pre-qualified range of shipping systems providing exceptional temperature control.
  • Pre-qualified 2 to 8 °C PCM shipper which delivers 3+ days or up to 82 hour performance
  • EPS insulation base and lid in robust outer corrugate
  • Universal packout so no need to manage the switch from summer to winter and back again
ChillTech® is a range of shipping systems providing exceptional temperature control for high value pharmaceuticals, biologics, or other temperature-sensitive products. Simple conditioning and mistake-proof packout ensure thatWięcej informacji o produkcie