Drukuj
Odważniki kalibracyjne, klasa E2, Proof Line
Dostawca: Sartorius Balances
YCW0121-02 YCW0221-02 YCW312-02 YCW512-02 YCW522-02 YCW422-02 YCW452-00 YCW612-00 YCW622-00 YCW452-02 YCW522-00 YCW612-02 YCW622-02 YCW412-00 YCW322-02 YCW352-00 YCW712-00 YCW1221-00 YCW2521-00 YCW2221-02 YCW1521-02 YCW0121-00 YCW2521-02 YCW1521-00 YCW2121-00 YCW2121-02 YCW2221-00 YCW0221-00 YCW412-02 YCW652-02 YCW422-00 YCW712-02 YCW352-02 YCW552-02 YCW312-00 YCW512-00 YCW322-00 YCW552-00 YCW652-00 YCW1121-00 YCW0521-00 YCW1221-AC-00 YCW312-AC-00 YCW2521-AC-02 YCW2521-AC-00 YCW2221-AC-02 YCW2221-AC-00 YCW2121-AC-02 YCW2121-AC-00 YCW1521-AC-02 YCW1221-AC-02 YCW1121-AC-02 YCW1121-AC-00 YCW0521-AC-02 YCW0521-AC-00 YCW0221-AC-02 YCW0221-AC-00 YCW0121-AC-02 YCW0121-AC-00 YCW1521-AC-00 YCW352-AC-02 YCW352-AC-00 YCW312-AC-02 YCW322-AC-02 YCW322-AC-00 YCW412-AC-00 YCW622-AC-02 YCW622-AC-00 YCW612-AC-02 YCW612-AC-00 YCW652-AC-00 YCW652-AC-02 YCW712-AC-02 YCW712-AC-00 YCW452-AC-02 YCW452-AC-00 YCW422-AC-02 YCW422-AC-00 YCW412-AC-02 YCW522-AC-00 YCW552-AC-02 YCW552-AC-00 YCW522-AC-02 YCW512-AC-02 YCW512-AC-00 YCW0521-02 YCW1121-02 YCW1221-02
SARBYCW0121-02EA 881 PLN
SARBYCW0121-02 SARBYCW0221-02 SARBYCW312-02 SARBYCW512-02 SARBYCW522-02 SARBYCW422-02 611-1703 SARBYCW612-00 SARBYCW622-00 SARBYCW452-02 611-1705 SARBYCW612-02 SARBYCW622-02 SARBYCW412-00 SARBYCW322-02 SARBYCW352-00 SARBYCW712-00 SARBYCW1221-00 SARBYCW2521-00 SARBYCW2221-02 SARBYCW1521-02 SARBYCW0121-00 SARBYCW2521-02 SARBYCW1521-00 SARBYCW2121-00 SARBYCW2121-02 SARBYCW2221-00 SARBYCW0221-00 SARBYCW412-02 SARBYCW652-02 SARBYCW422-00 SARBYCW712-02 SARBYCW352-02 SARBYCW552-02 SARBYCW312-00 611-1704 SARBYCW322-00 611-1706 611-1709 SARBYCW1121-00 SARBYCW0521-00 SARBYCW1221-AC-00 SARBYCW312-AC-00 SARBYCW2521-AC-02 SARBYCW2521-AC-00 SARBYCW2221-AC-02 SARBYCW2221-AC-00 SARBYCW2121-AC-02 SARBYCW2121-AC-00 SARBYCW1521-AC-02 SARBYCW1221-AC-02 SARBYCW1121-AC-02 SARBYCW1121-AC-00 SARBYCW0521-AC-02 SARBYCW0521-AC-00 SARBYCW0221-AC-02 SARBYCW0221-AC-00 SARBYCW0121-AC-02 SARBYCW0121-AC-00 SARBYCW1521-AC-00 SARBYCW352-AC-02 SARBYCW352-AC-00 SARBYCW312-AC-02 SARBYCW322-AC-02 SARBYCW322-AC-00 SARBYCW412-AC-00 SARBYCW622-AC-02 SARBYCW622-AC-00 SARBYCW612-AC-02 SARBYCW612-AC-00 SARBYCW652-AC-00 SARBYCW652-AC-02 SARBYCW712-AC-02 SARBYCW712-AC-00 SARBYCW452-AC-02 SARBYCW452-AC-00 SARBYCW422-AC-02 SARBYCW422-AC-00 SARBYCW412-AC-02 SARBYCW522-AC-00 SARBYCW552-AC-02 SARBYCW552-AC-00 SARBYCW522-AC-02 SARBYCW512-AC-02 SARBYCW512-AC-00 SARBYCW0521-02 SARBYCW1121-02 SARBYCW1221-02
Odważniki kalibracyjne, klasa E2, Proof Line
Odważniki Odważniki kalibracyjne
Sartorius Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • 0 to 5 mg leaf weights: Aluminium
  • 10 to 500 mg leaf weights: Nickel silver
  • 1 g to 10 Kg Knob weights: Stainless steel
Certyfikaty: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.