Drukuj

Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)

Dostawca: Rockland Immunochemicals
603-4302
ROCK603-4302EA 1600 PLN
ROCK603-4302
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
Przeciwciała
Secondary antibody reagents are ideal for western blotting, immunochemistry, ELISA, Fluorescence Microscopy, Flow Cytometry as well as other antibody detection methods.

Recommended Dilutions: ELISA: 1:150000; Western Blot: 1:2500 - 1:15000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR