Drukuj

Anti-HTT Rabbit Polyclonal Antibody

Dostawca: Rockland Immunochemicals
600-401-433
ROCK600-401-433EA 4120 PLN
ROCK600-401-433
Anti-HTT Rabbit Polyclonal Antibody
Przeciwciała
Anti-Huntingtin pS421 antibody has been tested for use in ELISA, immunochemistry and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect bands at approximately 350 kDa and 200 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:50 - 1:100;

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR