Drukuj

Anti-Dog serum Rabbit Polyclonal Antibody

Dostawca: Rockland Immunochemicals
104-4101
ROCK104-4101EA 2490 PLN
ROCK104-4101
Anti-Dog serum Rabbit Polyclonal Antibody
Przeciwciała
Anti-Canine serum is suitable for the specific deteciton of protiens found in dog serum. Cross reactivity to serum-protiens from other species has not been tested. Optimum experimental usage should be determined by researcher.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Dog serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR