Drukuj
Kolby filtrujące z gwintem, powlekane
Dostawca: ROBU GLASFILTER
34020 34500 34050
519-4141EA 638 PLN
519-4141 ROBU34500 ROBU34050
Kolby filtrujące z gwintem, powlekane
Kolby Kolby próżniowe
Filtering flasks, made of borosilicate glass 3.3. Highly resistant to most chemicals such as solvents, acids and alkalis.
  • Low pressure proof, sterilizable
  • With screw-on plastic hose
  • Protective coating