Drukuj
Zamrażarki skrzyniowe ULT, seria TDE, -86 °C
Dostawca: REVCO TECHNOLOGIES
TDEC06386FV TDEC25286FV TDEC39686FV
REVCTDEC06386FVEA 66290 PLN
REVCTDEC06386FV REVCTDEC25286FV REVCTDEC39686FV
Zamrażarki skrzyniowe ULT, seria TDE, -86 °C
Zamrażarki Zamrażarki niskotemperaturowe
Thermo Scientific™ TDE Series ultra-low temperature chest freezers set the standard for performance and reliable sample storage in heavy use situations. A sustainable solution with ultimate sample security and operational savings for every laboratory.
  • Improved performance
  • User friendliness and sustainability
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Three sizes (85, 360, and 566 L) to accommodate a variety of storage needs
  • F-Gas compliant with natural refrigerants and non-HFC
Więcej informacji o produkcie