Drukuj
Suszarka fluidalna, TG 200
Dostawca: Retsch GmbH
70.760.0001
RETS70.760.0001EA 50630 PLN
RETS70.760.0001
Suszarka fluidalna, TG 200
Eksykatory Suszarki ze złożem fluidalnym
The TG 200 fluid bed dryer is used in quality control, sample preparation and research departments. It allows the gentle drying of organic, inorganic, chemical or pharmaceutical bulk materials without localised overheating.
  • Gentle drying, dispersing and mixing also of temperature sensitive materials
  • Very short drying times (~ 5 to 20 min)
  • Powerful fan for optimal air throughput
  • Easy handling
  • Interval operation for better mixing of the fluidised bed
  • Wide range of accessories including various containers
  • Memory for up to 9 SOPs
  • Brushless motor provides long
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Przepływ 185 m³/h
Environmentally Preferable
Rozmiar podawanego materiału > 100 µm*
W×D×H 400×480×1000 mm
Weight 21 kg