Drukuj

Anti-CTDSP1 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
63-483
PRSI63-483EA 3110 PLN
PRSI63-483
Anti-CTDSP1 Rabbit Polyclonal Antibody
Przeciwciała
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: CTDSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 235-261
Host: Rabbit
Isotype: Ig
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR