Drukuj

Anti-STK11IP Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
59-923
PRSI59-923EA 3110 PLN
PRSI59-923
Anti-STK11IP Rabbit Polyclonal Antibody
Przeciwciała
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: STK11IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 385-411
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR