Drukuj

Anti-MBD3L3 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
56-236
PRSI56-236EA 3110 PLN
PRSI56-236
Anti-MBD3L3 Rabbit Polyclonal Antibody
Przeciwciała
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50 For IF starting dilution is: 1:10~50 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: MBD3L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR