Drukuj
Uvula Tissue Slide (Benign)
Dostawca: ProSci Inc.
13-003
PRSI13-003EA 3830 PLN
PRSI13-003
Uvula Tissue Slide (Benign)
Szkiełka mikroskopowe i nakrywkowe Microscopy Control Slides
Formalin-fixed, paraffin-embedded tissue is useful for a variety of applications including the targeting of proteins, RNA, and DNA by means of Immunohistochemistry (IHC), in-situ hybridization (ISH) and fluorescence in-situ hybridization (FISH), respectively.
Application Note: ProSci’s human tissue slides can be used with any antibody shown to be effective in formalin-fixed tissue using a standard immunostaining protocol. In order to avoid false negative results from DNA or RNA sequences not being present in a 4 µm cellular cross-section, 10 µm tissue sections are recommended. 10 µmWięcej informacji o produkcie