Drukuj

Anti-ZTS-00508841 Recombinant Antibody

Dostawca: ProSci Inc.
10-956
PRSI10-956EA 3280 PLN
PRSI10-956
Anti-ZTS-00508841 Recombinant Antibody
Przeciwciała
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR