Drukuj
Standards for water pollution QC check, mercury, VHG
Dostawca: LGC Standards PROMOCHEM
VHG-QWPHG-15
PROOVHG-QWPHG-15EA 454 PLN
PROOVHG-QWPHG-15
Standards for water pollution QC check, mercury, VHG
Standardy Wzorce do analiz środowiskowych
This solution is intended for use as a quality control check for methods related to mercury.
This standard has been preserved with approximately 1% (v/v) nitric acid and 0,04% (w/v) potassium dichromate. This standard has been prepared as a concentrate and must be diluted prior to analysis.
Certyfikaty: Traceable to NIST SRM.