Drukuj
Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Dostawca: LGC Standards PROMOCHEM
VHG-IPO4P-100 VHG-IPO4P-500
PROOVHG-IPO4P-100EA 238 PLN
PROOVHG-IPO4P-100 PROOVHG-IPO4P-500
Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Standardy Wzorce anionowe Phosphate(P) Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certyfikaty: Traceable to NIST and SRM.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable