Drukuj
Bufor transferowy Western Blot, 10X, bez metanolu, Pierce™
Dostawca: Thermo Fisher Scientific
35045
PIER35045EA 361 PLN
PIER35045
Bufor transferowy Western Blot, 10X, bez metanolu, Pierce™
Bufory Bufory do elektroforezy Bufory do transferu
Pierce™ Methanol-Free 10X Western Blot Transfer Buffer is a space-saving stock solution for preparing the methanol-free transfer buffer used with cooled, tank (wet) western blotting transfer apparatus.
  • Diluted 10-fold in water, the solution is ready to use for electrophoresis
  • No packets to open, no powder to dissolve, and no methanol required
  • Eliminates the possibility of powder remaining in a packet
  • Eliminates time spent waiting for powder to dissolve
  • Concentrated stock minimizes bench space needed for solutions
This concentrated Buffer is ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base.
The 10X Western Blot Transfer Buffer is ready to use upon dilution to 1X with water, since it does not contain methanol and the addition of methanol is not necessary for use in electrophoresis.