Drukuj
Kolumny i żele do usuwania endotoksyn, Detoxi-Gel™
Dostawca: Thermo Fisher Scientific
20339 20344 20340
PIER20339EA 1430 PLN
PIER20339 PIER20344 PIER20340
Kolumny i żele do usuwania endotoksyn, Detoxi-Gel™
Ekstrakcji białka Protein Extraction Kits
Detoxi-Gel™ endotoxin removing columns and gel uses polymixin B ligand immobilised on beaded affinity resin to bind and extract endotoxins from antibody or protein samples.
  • Resin uses immobilised polymixin B to remove pyrogens by binding their lipid A domains
  • 1 ml of gel binds 4000 to 6000 EU of the lipopolysaccharide from E. coli strain 055:BS
  • 1 ml of gel removes >9995 EU from a 10000 EU challenge of LPS (greater than 99% efficiency)
  • Reduces endotoxin levels in protein solutions, cell culture media, solutions containing pharmacological components and aqueous buffers
  • Stable and reusable resin can be regenerated by stripping off the endotoxins with a 1% deoxycholate solution in pyrogen-poor water